missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBNA1 Antibody (E1-2.5), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-312AF532
This item is not returnable.
View return policy
Description
EBNA1 Monoclonal specifically detects EBNA1 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| EBNA1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Virus | |
| IgG2b |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| E1-2.5 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| BKRF1, Epstein-Barr nuclear antigen 1, HHV4gp098, nuclear antigen EBNA1 | |
| Synthetic peptide corresponding to the repetitive Gly-Ala region of EBNA-1. | |
| 0.1 mL | |
| The antibody E1-2.5 recognizes repetitive Gly-Ala region of the Epstein-Barr virus (EBV) nuclear antigen 1 (EBNA-1). EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction