missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin Super Bright 645 Conjugate

Product Code. 16366102
Click to view available options
Quantity:
100 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16366102

Brand: Invitrogen™ 64431782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

eBioscience Streptavidin Super Bright 645 is a streptavidin-fluorochrome conjugate for indirect staining protocols to detect biotinylated primary antibodies in flow cytometry.

Super Bright dyes are polymer-based fluorophores that are excited by the violet laser (405 nm) and emit fluorescence at the wavelength indicated in their name. They are optimized for use in flow cytometry and allow for use of additional channels with violet excitation. Super Bright dyes are fully compatible with other commonly used fluorescent molecules, eBioscience buffers and fixatives, viability stains, and reagents.

Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.

Applications tested
Streptavidin Super Bright 645 Conjugate has been tested by flow cytometric analysis to detect biotinylated primary antibodies. The amount of this conjugate that will stain a cell sample in a final volume of 100 μL is less than or equal to 0.125 μg. Cell number should be determined empirically but can range from 105 to 108 cells/test. It is recommended that the streptavidin be carefully titrated for optimal performance in the assay of interest.

Samples can be stored in IC Fixation Buffer or 1-step Fix/Lyse Solution for up to three days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Clone specific performance after fixation should be determined empirically.

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Complete Staining Buffer for more information.

Super Bright polymer dyes are sold under license from Becton, Dickinson and Company.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/mL
Conjugate Super Bright 645
Quantity 100 μg
Content And Storage Store in refrigerator (2°C to 8°C).

Shipping Condition Wet Ice
Form Liquid
Product Line eBioscience™
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye Super Bright 645

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.