Learn More
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
Product Code. 15556286
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15556286 100 μL 100µL
15546286 4 x 100 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15556286 Supplier Invitrogen™ Supplier No. 00497793
  • ONLINE EXCLUSIVE PRICE
    £66.50
    £59.65 / 100µL
    Save £6.85
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer.
  • A ready to use solution of PHA-L in aqueous buffer
  • PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris
  • It is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity
  • This reagent is intended for use in in vitro activation of human and mouse leukocytes
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
Form Liquid
Product Line eBioscience
Product Type Phytohemagglutinin-L (PHA-L) Solution