missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Immune Response T Cell Cytokines Kit

Product Code. 17777473
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17777473 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17777473 Supplier Invitrogen™ Supplier No. A53421

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kit is comprised of a positive control stimulation reagent, protein transport inhibitors, buffers, viability dye, antibodies, and compensation beads needed to stimulate and detect production of Interferon gamma (IFNγ) and Interleukin 17A (IL-17A) from human T cells by flow cytometry.

  • Kit is comprised of a positive control stimulation reagent, protein transport inhibitors, buffers, viability dye, antibodies, and compensation beads
  • Use to stimulate and detect production of Interferon gamma (IFNγ) and Interleukin 17A (IL-17A) from human T cells by flow cytometry
  • Unique spectral fingerprint is designed to provide the brightest signal for flow cytometry experiments
  • Complete kit packaged in single-use vials for ease of use and to ensure stability performance over time
  • Extensive validation and protocol optimization for consistent results
  • Stimulation, fixation, permeabilization, and staining are performed in 96-well, round-bottom plates
  • Alternatively, cells may be cultured in bulk and transferred to 12 x 75 mm test tubes
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Antibodies, compensation beads, viability stain, buffers
Store at 4°C and –20°C.
For Use With (Application) T Cell Cytokine Analysis and Research
For Use With (Equipment) Flow Cytometry
Quantity 1 Kit
Regulatory Status RUO
Shipping Condition On Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.