missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Immune Response T Cell Activation Induced Markers Kit
Description
- Kit is comprised of a positive control stimulation reagent, buffers, viability dye, antibodies, and compensation beads
- Use to stimulate and detect expression of CD69, CD134 (OX-40), and CD137 (4-1BB) on human T cells by flow cytometry
- Unique spectral fingerprint is designed to provide the brightest signal for flow cytometry experiments
- Complete kit packaged in single-use vials for ease of use, and to ensure stability performance over time
- Extensive validation and protocol optimization for consistent results
- Stimulation, fixation, permeabilization, and staining are performed in 96-well, round-bottom plates
- Alternatively, cells may be cultured in bulk and transferred to 12 x 75 mm test tubes
Specifications
Specifications
| Content And Storage | Antibodies, compensation beads, viability stain, buffers Store at 4°C and –20°C. |
| For Use With (Application) | T Cell Activation Induced Marker Analysis and Research |
| For Use With (Equipment) | Flow Cytometry |
| Quantity | 1 kit |
| Regulatory Status | RUO |
| Shipping Condition | On Ice |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?