missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Concanavalin A (Con A) Solution (500X)
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
Pack of 4
Description
Con A is isolated from the Jack bean, Canavlia ensiformis. It is a lectin that binds alpha-D-glucose and alpha-D-mannose moieties found in various glycoproteins, glycolipids, and sugars and is a potent leukocyte mitogen. This reagent is intended for use in in vitro activation of human and mouse leukocytes.
Reported Application
Functional Assays
Specifications
Specifications
| Format | Vial |
| Quantity | 100 μL |
| Source | Canavlia Ensiformis |
| Form | Solution |
| Protein Subtype | Lectins |
| Product Line | eBioscience |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction