Learn More
Invitrogen™ eBioscience™ Avidin HRP
Description
Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.
This product is recommended for use in immunohistochemistry applications.
Conjugate: HRP
Reported Application: Immunohistochemical Staining
Specifications
Specifications
| Quantity | 100 μL |
| Target Molecule | Biotin |
| Content And Storage | Store at 2°C to 8°C |
| Formulation | Aqueous buffer, contains carrier protein/stabilizer, no sodium azide |
| Form | Liquid |
| For Use With (Application) | Immunohistochemical Staining |
| Product Line | eBioscience™ |
| Label or Dye | HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.