missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Avidin HRP

Product Code. 15548306
Click to view available options
Quantity:
100 μL
5 x 100 μL
Unit Size:
100µL
Pack of 5
This item is not returnable. View return policy

Product Code. 15548306

Brand: Invitrogen™ 18420089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies.

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.

This product is recommended for use in immunohistochemistry applications.

Conjugate: HRP

Reported Application: Immunohistochemical Staining

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 100 μL
Target Molecule Biotin
Content And Storage Store at 2°C to 8°C
Formulation Aqueous buffer, contains carrier protein/stabilizer, no sodium azide
For Use With (Application) Immunohistochemical Staining
Product Line eBioscience™
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.