missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBI3 Polyclonal Antibody

Description
A suggested positive control is rat spleen tissue lysate. PA5-20663 can be used with blocking peptide PEP-0783.
EBI3 (Epstein-Barr virus Induced-3) is a secreted glycoprotein of the hematopoietin receptor family. It plays a critical regulatory role in the induction of Th2-type immune responses and the development of Th2-mediated tissue inflammation in vivo, which may be mediated through the control of iNKT cell function. EBI3 dimerizes with p28 and p35 subunits of IL-12 to form new proteins IL-27 and IL-35, respectively. IL-27 is an early product of activated antigen presenting cell that is produced upon TLR ligation. It negatively regulates Th17 cell differentiation. EBI3 is widely expressed and its expression in dendritic cell is transcriptionally regulated by TLR signaling via MyD88 and NF-kappaB during innate immune responses preceding cytokine driven Th cell development. EBI3 signal inhibits delayed-type hypersensitivity responses by suppressing IL-17 production and inducing IL-10 hyperproduction. EBI3 may play a novel role in controlling tumor metastasis via lung CD8+ T cells, and its deficiency is associated with a diminished production of Th2 cytokines which are known to regulate allergic airway inflammation in asthma. Targeted deletion of EBI3 protects mice from lung metastasis.
Specifications
Specifications
| Antigen | EBI3 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | EBI3 |
| Gene Accession No. | O35228, Q14213 |
| Gene Alias | cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; IL-27; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; IL35 subunit; interleukin-27 subunit beta |
| Gene Symbols | EBI3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?