missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EBI3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818812
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818812 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818812 Supplier Invitrogen™ Supplier No. PA520662

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-20662 can be used with blocking peptide PEP-0782.

EBI3 (Epstein-Barr virus Induced-3) is a secreted glycoprotein of the hematopoietin receptor family. It plays a critical regulatory role in the induction of Th2-type immune responses and the development of Th2-mediated tissue inflammation in vivo, which may be mediated through the control of iNKT cell function. EBI3 dimerizes with p28 and p35 subunits of IL-12 to form new proteins IL-27 and IL-35, respectively. IL-27 is an early product of activated antigen presenting cell that is produced upon TLR ligation. It negatively regulates Th17 cell differentiation. EBI3 is widely expressed and its expression in dendritic cell is transcriptionally regulated by TLR signaling via MyD88 and NF-kappaB during innate immune responses preceding cytokine driven Th cell development. EBI3 signal inhibits delayed-type hypersensitivity responses by suppressing IL-17 production and inducing IL-10 hyperproduction. EBI3 may play a novel role in controlling tumor metastasis via lung CD8+ T cells, and its deficiency is associated with a diminished production of Th2 cytokines which are known to regulate allergic airway inflammation in asthma. Targeted deletion of EBI3 protects mice from lung metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBI3
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene EBI3
Gene Accession No. Q14213
Gene Alias cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; IL-27; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; IL35 subunit; interleukin-27 subunit beta
Gene Symbols EBI3
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human EBI3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10148
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.