missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Mouse anti-Human, Porcine, Rat, Clone: 1F3H10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | EBI3 |
|---|---|
| Clone | 1F3H10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828155
|
Proteintech
66354-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818155
|
Proteintech
66354-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBI3 is a subunit in two distinct heterodimeric cytokines: interleukin-27 (IL-27) and IL-35 (1-3). Like interleukin-23 (IL-23), IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. It exists as a heterodimer composed of the p40-related protein EBI3 and an IL-12 p35-related protein termed p28. IL-27 is produced after activation by antigen-presenting cells and induces proliferation of naive but not memory CD4+ T-cells. It acts by binding to its receptor WSX-1 (also known as TCCR) and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting the IL-27 is involved in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. IL-35 is composed of EBI3 and the p35 subunit of IL-12 and has been reported to have therapeutic effects against collagen-induced arthritis by expanding the population of regulatory T cells and suppressing Th17 cells. At least two isoform of EBI3 are known to exist.Specifications
| EBI3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EBI3, EBV induced gene 3 protein, Epstein Barr virus induced 3, IL 27 subunit beta, IL 27B, IL27B, Interleukin 27 subunit beta | |
| EBI3 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1F3H10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat, Human, Pig | |
| Q14213 | |
| 100522599, 10148, 680609 | |
| EBI3 Fusion Protein Ag3051 | |
| Primary | |
| -20°C | |
| EBI3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title