missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (12H5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26607-100ul
This item is not returnable.
View return policy
Description
EBI3 Monoclonal antibody specifically detects EBI3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| EBI3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 12H5 | |
| Western Blot 1:500-1:2000, ELISA | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| A synthesized peptide derived from Human EBI3 [UniProt Q14213] | |
| 100 μL | |
| Apoptosis, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction