missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104409
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EBF1.
Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. Target protein is a transcription factor that may regulate several olfactory-specific genes.
Specifications
| EBF1 | |
| Polyclonal | |
| Unconjugated | |
| EBF1 | |
| COE1; Collier; Collier, Olf and EBF transcription factor 1; early B cell factor 1; early B-cell factor; early B-cell factor (olfactory neuronal transcription factor 1); early B-cell factor 1; Ebf; EBF transcription factor 1; EBF1; O/E-1; OE-1; OLF1; Olf-1; Olfactory neuronal transcription factor; olfactory neuronal transcription factor 1; transcription factor COE1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1879 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UH73 | |
| EBF1 | |
| A synthesized peptide derived from human EBF1(Accession Q9UH73), corresponding to amino acid residues E398-S448. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction