missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebf1 Mouse anti-Mouse, PE, Clone: T26-818, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565494
This item is not returnable.
View return policy
Description
Ebf1 Monoclonal antibody specifically detects Ebf1 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| Ebf1 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| T26-818 | |
| PE | |
| Ebf1; Early B-cell factor 1; Ebf; Coe1; O/E-1; OE-1; Olf-1; Olf1 | |
| Mouse EBF1 Peptide | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction