missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ EBF-1 Antibody, Novus Biologicals™

Product Code. 18343958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343958 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343958 Supplier Novus Biologicals™ Supplier No. H00001879D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EBF-1 Polyclonal antibody specifically detects EBF-1 in Human samples. It is validated for Western Blot,ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBF-1
Applications Western Blot, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH41178.1
Gene Alias COE1, Collier, Olf and EBF transcription factor 1, Early B-cell factor, early B-cell factor 1, EBFFLJ41763, O/E-1, OE-1, OLF1FLJ39389, olfactory neuronal transcription factor 1, transcription factor COE1
Host Species Rabbit
Immunogen EBF1 (AAH41178.1, 1 a.a. - 560 a.a.) full-length human protein. MFGIQESIQRSGSSMKEEPLGSGMNAVRTWMQGAGVLDANTAAQSGVGLARAHFEKQPPSNLRKSNFFHFVLALYDRQGQPVEIERTAFVGFVEKEKEANSEKTNNGIHYRLQLLYSNGIRTEQDFYVRLIDSMTKQAIVYEGQDKNPEMCRVLLTHEIMCRFFLKFFLKCNQNCLKNAGNPRDMRRFQVVVSTTVNVDGHVLAVSDNMFVHNNSKHGRRARRLDPSEATPCIKAISPSEGWTTGGATVIIIGDNFFDGLQVIFGTMLVWSELITPHAIRVQTPPRHIPGVVEVTLSYKSKQFCKGTPGRFIYTALNEPTIDYGFQRLQKVIPRHPGDPERLPKEVILKRAADLVEALYGMPHNNQEIILKRAADIAEALYSVPRNHNQLPALANTSVHAGMMGVNSFSGQLAVNVSEASQATNQGFTRNSSSVSPHGYVPSTTPQQTNYNSVTTSMNGYGSAAMSNLGGSPTFLNGSAANSPYAIVPSSPTMASSTSLPSNCSSSSGIFSFSPANMVSAVKQKSAFAPVVRPQTSPPPTCTSTNGNSLQAISGMIVPPM
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1879
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.