missing translation for 'onlineSavingsMsg'
Learn More

EBE-A22, MedChemExpress

Product Code. 16438647
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438647

Brand: Medchem Express HY14347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EBE-A22 is a derivative of PD 153035 which can inhibit ErbB-1-phosphorylation, whereas EBE-A22 is inactive.

EBE-A22 is a derivative of PD 153035 which can inhibit ErbB-1-phosphorylation, whereas EBE-A22 is inactive.

Specifications

Chemical Name or Material EBE-A22
CAS 229476-53-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H16BrN3O2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (133.61 mM; Need ultrasonic)
SMILES BrC1=CC(N(C)C2=NC=NC3=CC(OC)=C(OC)C=C23)=CC=C1
Molecular Weight (g/mol) 374.23
Formula Weight 374.23
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.