missing translation for 'onlineSavingsMsg'
Learn More

EBE-A22, MedChemExpress

Product Code. 16482177
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482177 50 mg 50mg
16476446 100 mg 100mg
16476038 10 mg 10mg
16438647 5 mg 5mg
16430216 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16482177 Supplier Medchem Express Supplier No. HY14347
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress EBE-A22 is a derivative of PD 153035 which can inhibit ErbB-1-phosphorylation, whereas EBE-A22 is inactive.

EBE-A22 is a derivative of PD 153035 which can inhibit ErbB-1-phosphorylation, whereas EBE-A22 is inactive.

Specifications

Chemical Name or Material EBE-A22
CAS 229476-53-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H16BrN3O2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (133.61 mM; Need ultrasonic)
SMILES BrC1=CC(N(C)C2=NC=NC3=CC(OC)=C(OC)C=C23)=CC=C1
Molecular Weight (g/mol) 374.23
Formula Weight 374.23
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.