missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ebdarokimab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Ebdarokimab Humanized |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30282718
![]() |
Invitrogen™
MA559263 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30282380
|
Invitrogen™
MA559264 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ebdarokimab, also known as AK101, is a monoclonal antibody that targets interleukin-12 (IL-12) and interleukin-23 (IL-23), two cytokines implicated in the inflammatory response associated with plaque psoriasis. Interleukin-12 subunit beta (IL-12b), also known as IL-12p40, is a crucial component of the heterodimeric cytokine IL-12 and IL-23. The IL-12b gene is located on chromosome 5q31-33 and encodes for a 40 kDa protein. IL-12 is composed of two subunits, p35 and p40, while IL-23 is composed of p19 and the shared p40 subunit. The IL-12b protein has a four-helix bundle structure, homologous to other members of the hematopoietin superfamily. Functionally, IL-12b plays a significant role in immune responses by promoting the differentiation of naive T cells into Th1 cells and enhancing the cytotoxic activity of natural killer (NK) cells. It is essential in the immune system's response to intracellular pathogens. Dysregulation of IL-12b has been implicated in various autoimmune diseases and cancers, making it a target for therapeutic interventions.Specifications
| Ebdarokimab Humanized | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title