missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebastine, MedChemExpress
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
Description
Ebastine (LAS-W 090) is an orally active, second-generation histamine H1 receptor antagonist. Ebastine can be used for the symptoms of allergic rhinitis and chronic idiopathic urticaria research.
Specifications
Specifications
| Chemical Name or Material | Ebastine |
| Physical Form | Solid |
| CAS | 90729-43-4 |
| Color | White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C32H39NO2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | LAS-W 090 RP64305 |
| Solubility Information | Ethanol : 58.75 mg/mL (125.09 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (17.74 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction