missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebastine, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ebastine |
| CAS | 90729-43-4 |
| Analyte or Component Names | Ebastine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32 H39 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ebastine, 1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-piperidinyl]-1-butanone, Bastel, Ebastel, Ebastin, LAS-W 090, RP 64305 |
| InChI Formula | InChI=1S/C32H39NO2/c1-32(2,3)28-18-16-25(17-19-28)30(34)15-10-22-33-23-20-29(21-24-33)35-31(26-11-6-4-7-12-26)27-13-8-5-9-14-27/h4-9,11-14,16-19,29,31H,10,15,20-24H2,1-3H3 |
| Solution Type | Neat |
| SMILES | CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)OC(c3ccccc3)c4ccccc4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction