missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBAG9/RCAS1 Mouse anti-Human, Clone: CPTC-EBAG9-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07534-100ug
This item is not returnable.
View return policy
Description
EBAG9/RCAS1 Monoclonal specifically detects EBAG9/RCAS1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| EBAG9/RCAS1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
| Recombinant human full-length EBAG9/RCAS1 protein | |
| 100 μg | |
| Apoptosis | |
| 9166 | |
| Store at 4°C. | |
| IgG2c κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CPTC-EBAG9-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction