missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB2 Antibody (OTI1F3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71650JF646
This item is not returnable.
View return policy
Description
EB2 Monoclonal specifically detects EB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| EB2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAPRE2 | |
| Full length recombinant protein of human EB2 (NP_055083) produced in HEK293T cells. | |
| 0.1 mL | |
| 10982.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1F3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| APC-binding protein EB2, EB1, EB2APC-binding protein EB1, End-binding protein 2, microtubule-associated protein, RP/EB family, member 2, RP1microtubule-associated protein RP/EB family member 2, T-cell activation protein, EB1 family | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction