missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582107
This item is not returnable.
View return policy
Description
Immunogen sequence: VAPSLVAPAL NKPKKPLTSS SAAPQRPIST QRTAAAPKAG PGVVRKNPGV GNGDDEAAEL MQQVNVLKLT VEDLEKERDF YFGKLRNIEL ICQENEGEND PVLQRIVDIL YATDEGFVIP DE Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 93%.
EB1 family proteins are evolutionarily conserved proteins that bind microtubule plus-ends and centrosomes and regulate the dynamics and organization of microtubules. The MAPRE genes encode the EB1 family proteins. The human MAPRE genes MAPRE1, MAPRE2, and MAPRE3 code for EB1, RP1, and EBF3 proteins, respectively. EB1 family members appear to be expressed ubiquitously. The EB1 (MAPRE1) gene, at 20q11. 2, is fused to MLL in an adult patient with pro-B acute lymphoblastic leukemia. EB1 is a microtubule-associated protein that interacts with the colorectal adenomatous polyposis coli tumor-suppressor protein and plays important roles in regulating microtubule dynamics, cell polarity, and chromosome stability.
Specifications
| EB1 | |
| Polyclonal | |
| Unconjugated | |
| Mapre1 | |
| 5530600P05Rik; adenomatosis polyposis coli binding protein Eb1; adenomatous polyposis coli-binding protein EB1; AI462499; AI504412; APC-binding protein EB1; AW260097; BIM1p; D2Ertd459e; Eb1; end-binding protein 1; Mapre1; MGC117374; MGC129946; microtubule associated protein RP/EB family member 1; Microtubule-associated protein RP/EB family member 1; microtubule-associated protein RP/EB family member 1-like protein; microtubule-associated protein, RP/EB family, member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114764, 13589, 22919 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15691, Q61166, Q66HR2 | |
| Mapre1 | |
| Recombinant protein corresponding to Human MAPRE1. Recombinant protein control fragment (Product #RP-102164). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction