Learn More
Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps, Sustain™ Series
Product Code. 30179688 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 Strips
Unit Size:
Pack of 250
Product Code. Quantity Unit Size
30179688 250 Strips Pack of 250
1 options
Product Code. 30179688 Supplier Thermo Scientific™ Supplier No. AB2000SS
  • ONLINE EXCLUSIVE PRICE
    £323.00
    £290.65 / Pack of 250
    Save £32.35
    10% Off
View all promotions
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Thermo Scientific™ EasyStrip Plus Tube Strip with Attached Flat Caps, Sustain Series, is designed to reduce the risk of cross-contamination in PCR assays.
The Thermo Scientific™ EasyStrip Plus Tube Strip with Attached Flat Caps, Sustain Series, is designed to reduce the risk of cross-contamination in PCR assays. The tube strip is compatible with 0.2 mL thermal cycler blocks and features individually attached flat caps for quick and easy access to individual samples. In addition, graduated 20 μL markers on each tube enable visual volume verification and measurement of reagent and sample additions.
Features and benefits
  • Suitable for PCR applications
  • Avoid cross contamination with the individually attached caps
  • Verify tube reaction volumes with graduated 20 μL markings on each tube
  • Track samples using the end tabs to label each strip
  • Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
  • Certified DNase-, RNase-, human DNA-, and PCR inhibitor-free
A breakthrough in sustainable PCR technology
Thermo Scientific™ Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific™ PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
  • Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
  • Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
  • Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific™ PCR plastics
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Color Natural
Quantity 250 Strips
Material Polypropylene
Closure Type Flat Cap (Attached)
Autoclavable Autoclavable
Closure Included Yes
Green Features ISCC PLUS