Learn More
STEMCELL Technologies EasyPlate™ EasySep™ Magnet
Product Code. 17117851
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 107 Cells (mouse splenocytes) or 1 x 107 Cells (human PBMC)/well
Unit Size:
1 set
Product Code. Quantity Unit Size
17117851 2 x 107 Cells (mouse splenocytes) or 1 x 107 Cells (human PBMC)/well 1 set
1 options
Product Code. 17117851 Supplier STEMCELL Technologies Supplier No. 18102
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Immunomagnetic column-free magnet
  • The EasyPlate™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a small number of cells (up to 2 x 10^7 cells per well)
  • The EasyPlate™ EasySep™ Magnet generates a high-gradient magnetic field that is strong enough to separate cells labeled with EasySep™ D Magnetic Particles without the use of columns
  • The EasyPlate™ EasySep™ Magnet is designed to hold a round-bottom non-tissue culture-treated 96-well plate
Specifications
For Use With (Application) Cell Isolation
Quantity 2 x 107 Cells (mouse splenocytes) or 1 x 107 Cells (human PBMC)/well