Learn More
Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters
Use this easy-to-use sprayer in your sensitive LC-MS separation of protein and peptides using nano and capillary LC.
Brand: Thermo Scientific™ ES791
Code : 15
Additional Details : Weight : 0.10000kg
Description
Connect your nano or capillary column to your Thermo Scientific™ mass spectrometers (MS) more easily using Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters. The nano and capillary emitters act as a column-independent sprayer, allowing the introduction of low flow from nano and capillary columns without troublesome handling of traditional spray needles. There are two versions of emitters available: with and without integrated transfer lines. These sprayers are ideal for protein and peptide separation using nano and capillary HPLC.
- Available in nanoflow and microflow formats to meet a wider range of tuning requirements
- Thermo Scientific™ Dionex™ nanoViper™ fitting makes installation easy
- Precision positioned glass emitter for excellent spray performance
- Integrated zero-dead-volume unions ensure reliability
Specifications
EASY-Spray Emitter | |
500 mm | |
Unpacked | |
EASY-Spray Emitter with transfer line |
0.02 mm | |
Easy Spray | |
Emitter |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.