Learn More
Vici Valco Instruments™ Easy-Flange Kit
Product Code. 12649059
Change view
Click to view available options
For Use With:
1/16 in. to 1/8 in. OD PTFE Tubing
Unit Size:
Each
Product Code. For Use With Unit Size
12649059 1/16 in. to 1/8 in. OD PTFE Tubing Each
1 options
Product Code. 12649059 Supplier Vici Valco Instruments™ Supplier No. JR201540
  • ONLINE EXCLUSIVE PRICE
    £625.25
    £154.96 / Pack of 1
    Save £470.29
    75% Off
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Flange-rolling tool that uses mechanical force to create flanges in PTFE and PTFE-like tubes. Vici Valco Instruments™ Easy-Flange Kit does not require any electrical power.
  • Flange formation in PTFE and PTFE-like tubing without electrical power
  • Suitable for tubes with OD between 1/16 in. and 1/8 in.
  • Flange quality is improved because flanges are formed without applying heat stress
  • Specially designed negative conical profile of the flange forming part provides an ideal shape for maximum sealing properties
Specifications
Type Easy-Flange Kit
Includes Plastic box, Flanging discs with: 0.5 mm Stainless Steel pin for PEEK tubing, 0.8 mm Polymer pin, 0.8 mm Titanium pin, 1.3 mm Polymer pin, 1.3 mm Titanium pin, Clean-cut tubing cutter, PTFE tubing (1/16 in. x 0.75 mm ID, 3 m/Pk)
For Use With 1/16 in. to 1/8 in. OD PTFE Tubing