Learn More
Thermo Scientific™ EASY-Column™ Capillary HPLC Column Connector Kits
Product Code. 11571174 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
A/B mixing and venting for two-column setup
HPLC to column
Kit for 360μm OD fused silica
Two-column setup
Unit Size:
Each
Pack of 30
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11571174 A/B mixing and venting for two-column setup Each
11875372 Kit for 360μm OD fused silica Pack of 30
11322150 Two-column setup Each
11302150 HPLC to column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11571174 Supplier Thermo Scientific™ Supplier No. SC601
Only null left
Add to basket
Add to basket
Use these connectors, tees and sleeves with Thermo Scientific™ EASY-Column™ Capillary HPLC Columns.
Use these connector kits, sleeves, and tees with Thermo Scientific™ EASY-Column™ Capillary Columns. EASY-Columns optimize nanoscale LC and LC/MS with. Easy-to-use, zero dead volume connector kits are available for one- and two-column configurations and ensure simple, reproducible setup for on-line LC/MS. Each column delivers excellent chromatographic performance on any nanoscale LC system. At flow rates of nL/min versus mL/min, the EASY-Column offers higher sensitivity with greater signal-to-noise ratio compared to traditional electrospray.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.79 mm
For Use With LC/MS
Pore Size 120 Å
Product Line EASY-Column
Endcapped Yes
Type A/B mixing and venting for two-column setup
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.