missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Easy Breeze™ Gel Dryer Accessories: Frame Components and Accessories

Product Code. 11505604 Shop All Cytiva Products
Change view
Click to view available options
Type:
Quarter-turn Knob
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11505604 Quarter-turn Knob Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11505604 Supplier Hoefer™ Supplier No. SE1213

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with the SE 1200 Easy Breeze Gel Drying System

  • Highly resistant to acids, bases, and all commonly used buffer systems
  • Easily removed while maintaining gel integrity
  • Lightweight and virtually unbreakable when used with reasonable care
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) SE 1200 Easy Breeze Gel Drying System
Type Quarter-turn Knob
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.