Learn More
Abeba™ Easy 7630 Shoes
Product Code. 15692908
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15692908
Brand: Abeba™ 7630/44
Only null left
Add to basket
Add to basket
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • White
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Polyurethane
Style 7630