Learn More
Abeba™ Easy 37623 Shoes
Product Code. 15691108
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
Unit Size:
1 pair
Product Code. 15691108
Brand: Abeba™ 37623/37
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/red
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Size 37
Sole Features Slip-resistant
Sole Material Polyurethane
Style 37623