Learn More
Scie Plas Easigel H1-SET Horizontal Gel Unit
For standard preparative and analytical studies of nucleic acids
Brand:  Scie Plas H1-SET
Additional Details : Weight : 0.75200kg
Product Code. 15825571
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Plastic casting gates
  • Colour-coded combs
  • Quick-release lid to disconnect tank and lid simultaneously
  • UV-transparent acrylic, for direct visualisation of gel within tank
  • Compact design providing optimal control over voltage gradient and run-time
  • Small size. allows safe and easy transportation
Specifications
Specifications
Gel Unit
Easigel horizontal gel unit with built-in casting tray and2 x 1.5mm thick, 8-sample combs
50 mL
135 mm
70 to 90 V
80 x 100 mm
40
8 x 10 cm (gel)
100 mm