Learn More
Invitrogen™ EARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113954
Description
Immunogen sequence: TAKHLLLYQA LGWQPPHFAH LPLLLNRDGS KLSKRQGDVF LEHFAADGFL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%; Yeast: 91%; Zebrafish: 93%.
Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the minimal set of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
Specifications
| EARS2 | |
| Polyclonal | |
| Unconjugated | |
| Ears2 | |
| 3230401I01Rik; COXPD12; Ears2; GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; glutamyl-tRNA synthetase 2 (mitochondrial)(putative); glutamyl-tRNA synthetase 2 mitochondrial (putative); glutamyl-tRNA synthetase 2, mitochondrial; Kiaa1970; mKIAA1970; MSE1; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial; RGD1307904 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 124454 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5JPH6 | |
| Ears2 | |
| Synthetic peptide directed towards the middle region of human EARS2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.