missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EARS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15718281 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15718281 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15718281 Supplier Invitrogen™ Supplier No. PA560445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YQGSFILRLE DTDQTRVVPG AAENIEDMLE WAGIPPDESP RRGGPAGPYQ QSQRLELYAQ ATEALLKTGA AYPCFCSPQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the minimal set of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EARS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ears2
Gene Accession No. Q5JPH6
Gene Alias 3230401I01Rik; COXPD12; Ears2; GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; glutamyl-tRNA synthetase 2 (mitochondrial)(putative); glutamyl-tRNA synthetase 2 mitochondrial (putative); glutamyl-tRNA synthetase 2, mitochondrial; Kiaa1970; mKIAA1970; MSE1; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial; RGD1307904
Gene Symbols Ears2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EARS2. Recombinant protein control fragment (Product #RP-98631).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124454
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.