missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Earplugs Zero Waste Box™

Product Code. 17855905
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17855905 Medium -
17845815 Small Each
17835705 Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17855905 Supplier Terracycle™ Supplier No. CS44MUK
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Earplugs using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Foam and plastic earplugs

Approximate waste units accepted:

  • Small: 880 earplugs
  • Medium: 2400 earplugs
  • Large: 4400 earplugs

Do not send in:

  • Wax earplugs
  • Hairnets, gloves, lab coats or other garments
  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters and broken glass

Specifications

Capacity (Metric) 15 kg
Capacity (English) 33 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 30.5 x 30.5 x 84 cm
Disposable Single-use
Latex Free Yes
Size Medium
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.