Learn More
Sperian™ Howard Leight™ Bilsom™ 303 Disposable Earplugs
Product Code. 15221149
Change view
Click to view available options
Color: Yellow and White
Product Code.
15221149 Color: Yellow and White
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15221149 Supplier Sperian™ Supplier No. 1006187
  • ONLINE EXCLUSIVE PRICE
    £26.75
    £25.93 / Pack of 10
    Save £0.82
    3% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Wears more comfortably than other foam earplugs over long periods of time. Sperian™ Howard Leight™ Bilsom™ 303 Disposable Earplugs are ideal for medium- to high-noise environments.
  • Polyurethane foam expands gently in ear canal, resulting in less expansion pressure
  • Smooth, soil-resistant skin prevents dirt build-up
  • Tapered bullet-shaped design is easier to roll-down and insert, adapts more evenly to ear canal
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, White
Size Universal
Quantity 1 Ea.
Plug Shape Bullet
Product Type Earplug