missing translation for 'onlineSavingsMsg'
Learn More

EAR2/NR2F6 Antibody (EM-51) - BSA Free, Novus Biologicals™

Product Code. 18335123 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335123 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335123 Supplier Novus Biologicals Supplier No. NBP309110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EAR2/NR2F6 Monoclonal specifically detects EAR2/NR2F6 in Human samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAR2/NR2F6
Applications Western Blot, Flow Cytometry, Flow Cytometry
Classification Monoclonal
Clone EM-51
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 3-9 ug/ml, Flow (Intracellular) 3-9 ug/ml
Formulation PBS
Gene Alias EAR-2ERBA-related gene-2, EAR2nuclear receptor subfamily 2 group F member 6, ERBAL2v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2, nuclear receptor subfamily 2, group F, member 6, V-erbA-related protein 2
Host Species Mouse
Immunogen Recombinant human EAR2/NR2F6 protein expressed in E. coli
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2063
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.