Learn More
Sperian™ Honeywell™ AirSoft™ Red Vinyl Cord Earplugs
Product Code. 15369771
Product Code. 15369771 Supplier Sperian™ Supplier No. 1030612
Only null left
Add to basket
Add to basket
With four soft flanges to offer a better fit in the ear canal for a tighter seal and better attenuation. Sperian™ Honeywell™ AirSoft™ Red Vinyl Cord Earplugs deliver superior long-term comfort.
  • Patented soft shell around a pocket of air
  • Distinctive blue and red colors aid in visibility
  • Easy clean-up with soap and water allows for weeks of continuous use
TRUSTED_SUSTAINABILITY
Specifications
Color Blue, Red
Size Universal
Plug Shape Flanged