missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT3 Antibody (S180-41), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59319
This item is not returnable.
View return policy
Description
EAAT3 Monoclonal antibody specifically detects EAAT3 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
| EAAT3 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| SLC1A1 | |
| Fusion protein amino acids 1-524 (full length) of human EAAT3 (Uniprot: P43005) | |
| 100 μg | |
| Primary | |
| Detects 60kDa. | |
| Store at -20C. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| S180-41 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6505 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction