missing translation for 'onlineSavingsMsg'
Learn More

EAAT3 Antibody (4D6.2), DyLight 650, Novus Biologicals™

Product Code. 18418585 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418585 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418585 Supplier Novus Biologicals Supplier No. NBP229747C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EAAT3 Monoclonal antibody specifically detects EAAT3 in Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAAT3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 4D6.2
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1
Host Species Mouse
Immunogen C-terminus peptide (KDKSDTISFTQTSQF).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 6505
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.