missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22515-1-AP-150UL
This item is not returnable.
View return policy
Description
Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.Specifications
| EAAT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC1A2 | |
| EAAT2, GLT 1, GLT1, SLC1A2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20511, 29482, 6506 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31596, P43004, P43006 | |
| SLC1A2 | |
| EAAT2 Fusion Protein Ag18247 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction