missing translation for 'onlineSavingsMsg'
Learn More

E7449, MedChemExpress

Product Code. p-200101766
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483296

Brand: Medchem Express HY12418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E7449 is a potent PARP1 and PARP2 inhibitor and also inhibits TNKS1 and TNKS2, with IC50s of 2.0, 1.0, ∼50 and ∼50 nM for PARP1, PARP2, TNKS1 and TNKS2, respectively, using 32P-NAD+ as substrate.

E7449 is a potent PARP1 and PARP2 inhibitor and also inhibits TNKS1 and TNKS2, with IC50s of 2.0, 1.0, ∼50 and ∼50 nM for PARP1, PARP2, TNKS1 and TNKS2, respectively, using 32P-NAD+ as substrate.

Specifications

Chemical Name or Material E7449
Physical Form Solid
CAS 1140964-99-3
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C18H15N5O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 6.4 mg/mL (20.17 mM; Need ultrasonic and warming)
SMILES O=C1NNC2=C3C1=CC=CC3=NC(CN4CC5=C(C=CC=C5)C4)=N2
Molecular Weight (g/mol) 317.34
Formula Weight 317.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.