missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E7449, MedChemExpress
Description
E7449 is a potent PARP1 and PARP2 inhibitor and also inhibits TNKS1 and TNKS2, with IC50s of 2.0, 1.0, ∼50 and ∼50 nM for PARP1, PARP2, TNKS1 and TNKS2, respectively, using 32P-NAD+ as substrate.
Specifications
Specifications
| Chemical Name or Material | E7449 |
| Physical Form | Solid |
| CAS | 1140964-99-3 |
| Color | Yellow |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H15N5O |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 6.4 mg/mL (20.17 mM; Need ultrasonic and warming) |
| SMILES | O=C1NNC2=C3C1=CC=CC3=NC(CN4CC5=C(C=CC=C5)C4)=N2 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?