missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E74 like factor 1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1069
This item is not returnable.
View return policy
Description
E74 like factor 1 Polyclonal specifically detects E74 like factor 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| E74 like factor 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| E74-like factor 1, E74-like factor 1 (ets domain transcription factor), ETS-related transcription factor Elf-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1997 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.03 - 0.1 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_758961. | |
| ELF1 | |
| Peptide with sequence C-AMKQNELLEPNSF corresponding to C-Terminus according to NP_758961.1, NP_001138825.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction