missing translation for 'onlineSavingsMsg'
Learn More

E6AP/UBE3A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18351394 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351394 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351394 Supplier Novus Biologicals Supplier No. NBP298953AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

E6AP/UBE3A Polyclonal specifically detects E6AP/UBE3A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E6AP/UBE3A
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human E6AP/UBE3A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7337
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.