Learn More
Invitrogen™ E6AP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595918
Description
Immunogen sequence: KLIRLWSKYN ADQIRRMMET FQQLITYKVI SNEFNSRNLV NDDDAIVAAS KCLKMVYYAN VVGGEVDTNH NEEDDEEPIP ESSELTLQEL LGEERRNKKG PRVDPLETEL GVKTLDCRKP LIPFEEFINE PLNEVLEMDK DYTFFKVETE NKFSFMTCPF ILNAVTKNLG LYYDNRIRMY SERRITVLYS LVQGQQLNPY LRLKVRRDHI IDDALVRLEM IAMENPADLK KQLYVEFEGE QGVDEGGVSK EFFQLVVEEI FNPDIGMFTY DESTKLFWFN PSSFETEGQF T; Positive Samples: 293T, SKOV3, BT-474, U-251MG, Jurkat, Mouse brain, Mouse heart, Mouse thymus, Rat brain; Cellular Location: Cytoplasm, Nucleus.
UBE3A (ubiquitin-protein ligase E3A, AKA E6AP) is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and transfers it to its substrates. Several substrates have been identified including the ARNTL/BMAL1, ARC, RAD23A, RAD23B, MCM7, annexin A1, the PML tumor suppressor, and the cell cycle regulator CDKN1B. Additionally, UBE3A may function as a cellular quality control ubiquitin ligase by helping the degradation of the cytoplasmic misfolded proteins. UBE3A also plays an important role in the regulation of the circadian clock. Defects in UBE3A are a cause of Angelman syndrome (AS).
Specifications
| E6AP | |
| Polyclonal | |
| Unconjugated | |
| UBE3A | |
| 4732496B02; 5830462N02Rik; A130086L21Rik; ANCR; AS; CTCL tumor antigen se37-2; E6AP; E6-AP; E6-AP ubiquitin protein ligase; E6AP ubiquitin-protein ligase; EPVE6AP; FLJ26981; HECT-type ubiquitin transferase E3A; HPVE6A; human papilloma virus E6-associated protein; human papillomavirus E6-associated protein; mKIAA4216; oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54; UBE3A; ubiquitin conjugating enzyme E3A; ubiquitin protein ligase E3A; ubiquitin-protein ligase E3A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22215, 7337 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08759, Q05086 | |
| UBE3A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-590 of human UBE3A (NP_5708531). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.