missing translation for 'onlineSavingsMsg'
Learn More

E4CPG, MedChemExpress

Product Code. 16494286
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494286

Brand: Medchem Express HY100372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E4CPG ((RS)-ECPG) is a Group I/Group II metabotropic glutamate receptor (mGluR) antagonist. E4CPG can inhibit the paired-pulse ratio of monosynaptic inhibitory postsynaptic currents (IPSC) potentiation.

E4CPG ((RS)-ECPG) is a Group I/Group II metabotropic glutamate receptor (mGluR) antagonist. E4CPG can inhibit the paired-pulse ratio of monosynaptic inhibitory postsynaptic currents (IPSC) potentiation.

Specifications

Chemical Name or Material E4CPG
Physical Form Solid
CAS 170846-89-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H13NO4
For Use With (Application) Neuroscience-Neuromodulation
Synonym (RS)-ECPG
Solubility Information DMSO : 10 mg/mL (44.80 mM; ultrasonic and adjust pH to 12 with NaOH) ∣H2O : 1 mg/mL (4.48 mM; ultrasonic and warming and heat to 80°C)
SMILES NC(C(O)=O)(CC)C1=CC=C(C(O)=O)C=C1
Molecular Weight (g/mol) 223.23
Formula Weight 223.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.