missing translation for 'onlineSavingsMsg'
Learn More

E3 ligase Ligand 8, MedChemExpress

Product Code. 16481929
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481929 1 g 1g
16450346 2 g 2g
16441916 500 mg 500mg
16437836 100 mg 100mg
16423977 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16481929 Supplier Medchem Express Supplier No. HY43961
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress E3 ligase Ligand 8 is a ligand for E3 ubiquitin ligase. E3 ligase Ligand 8 can be connected to the ligand for protein by a linker to form PROTACs. PROTACs are inducers of ubiquitination-mediated degradation of cancer-promoting proteins.

E3 ligase Ligand 8 is a ligand for E3 ubiquitin ligase. E3 ligase Ligand 8 can be connected to the ligand for protein by a linker to form PROTACs. PROTACs are inducers of ubiquitination-mediated degradation of cancer-promoting proteins.

Specifications

Chemical Name or Material E3 ligase Ligand 8
CAS 1225383-33-4
Color White
Purity Grade Notes Research
Molecular Formula C31H34N2O6
Quantity 1 g
Solubility Information DMSO : 100 mg/mL (188.46 mM; Need ultrasonic)
SMILES CC(C)C[C@@H](C(O)=O)NC([C@@H](O)[C@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC4=CC=CC=C4)=O
Molecular Weight (g/mol) 530.61
Formula Weight 530.61
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.