missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529479
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HepG2. Predicted reactivity: Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation (Christensen et al., 2005).
Specifications
| E2F8 | |
| Polyclonal | |
| Unconjugated | |
| E2F8 | |
| 4432406C08Rik; AA410048; E2F 8; E2F family member 8; E2F transcription factor 8; E2F8; E2F-8; FLJ23311; RGD1308091; Transcription factor E2F8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| A0AVK6 | |
| E2F8 | |
| Recombinant protein encompassing a sequence within the center region of human E2F8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction