missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F7 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52648AF647
This item is not returnable.
View return policy
Description
E2F7 Polyclonal antibody specifically detects E2F7 in Human samples. It is validated for Western Blot
Specifications
| E2F7 | |
| Polyclonal | |
| Western Blot | |
| E2F transcription factor 7, E2F-7, FLJ12981, transcription factor E2F7 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human E2F Transcription Factor 7 using the numbering given in entry NP_976328.2 (GeneID 144455). | |
| 0.1 mL | |
| Signal Transduction | |
| 144455 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction